N-(4-[1,3]Dioxolan-2-yl-pyridin-3-yl)-2,2-dimethylpropionamide

Supplier: Matrix Scientific

026143-500MG
101821-688EA 315.38 USD
101821-688
N-(4-[1,3]Dioxolan-2-yl-pyridin-3-yl)-2,2-dimethylpropionamide
N-(4-[1,3]Dioxolan-2-yl-pyridin-3-yl)-2,2-dimethylpropionamide

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₃
MW: 250.3 g/mol
MDL Number: MFCD07776835
CAS Number: 869735-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR