N-(5-Iodo-6-methyl-pyridin-2-yl)-2,2-dimethylpropionamide

Supplier: Matrix Scientific

026139-500MG
101821-682EA 250.77 USD
101821-682
N-(5-Iodo-6-methyl-pyridin-2-yl)-2,2-dimethylpropionamide
N-(5-Iodo-6-methyl-pyridin-2-yl)-2,2-dimethylpropionamide

Hazard: Irritant

Formula: C₁₁H₁₅IN₂O
MW: 318.16 g/mol
MDL Number: MFCD04150999
CAS Number: 676588-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR