N-(3-Hydroxypyridin-4-yl)-2,2-dimethylpropionamide

Supplier: Matrix Scientific

026136-500MG
101821-676EA 315.38 USD
101821-676
N-(3-Hydroxypyridin-4-yl)-2,2-dimethylpropionamide
N-(3-Hydroxypyridin-4-yl)-2,2-dimethylpropionamide

Hazard: Irritant

Formula: C₁₀H₁₄N₂O₂
MW: 194.24 g/mol
MDL Number: MFCD07776841
CAS Number: 169205-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR