(4-Aminopyridin-3-yl)methanol
Supplier: Matrix Scientific
026134-1G
101821-672EA
180
USD
101821-672
(4-Aminopyridin-3-yl)methanol
(4-Aminopyridin-3-yl)methanol
Hazard: Irritant
Formula:
C₆H₈N₂O |
MDL Number:
MFCD06203066 CAS Number: 138116-34-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...