5-Bromo-2-chloro-3-iodopyridine

Supplier: Matrix Scientific

Synonyms: 5-Bromo-2-chloro-3-iodo-pyridine

026129-250MG
101821-662EA 15.38 USD
101821-662
5-Bromo-2-chloro-3-iodopyridine
5-Bromo-2-chloro-3-iodopyridine

Hazard: Irritant

Formula: C₅H₂BrClIN
MW: 318.34 g/mol
MDL Number: MFCD08688585
CAS Number: 928653-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR