5-Bromo-3-iodo-2-isopropoxypyridine

Supplier: Matrix Scientific

026127-500MG
101821-658EA 250.77 USD
101821-658
5-Bromo-3-iodo-2-isopropoxypyridine
5-Bromo-3-iodo-2-isopropoxypyridine

Hazard: Irritant

Formula: C₈H₉BrINO
MW: 341.98 g/mol
MDL Number: MFCD07366719
CAS Number: 848243-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR