3-(2,2-Dimethylpropionylamino)isonicotinic acid

Supplier: Matrix Scientific

Synonyms: 3-(2,2-Dimethylpropanamido)pyridine-4-barboxylic acid

026122-500MG
101821-648EA 160 USD
101821-648
3-(2,2-Dimethylpropionylamino)isonicotinic acid
3-(2,2-Dimethylpropionylamino)isonicotinic acid

Hazard: Irritant

Formula: C₁₁H₁₄N₂O₃
MW: 222.25 g/mol
MDL Number: MFCD06659008
CAS Number: 86847-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR