N-(4-tert-Butylsulfanylpyridin-3-yl)-2,2-dimethyl-propionamide

Supplier: Matrix Scientific

026118-500MG
101821-638EA 258.46 USD
101821-638
N-(4-tert-Butylsulfanylpyridin-3-yl)-2,2-dimethyl-propionamide
N-(4-tert-Butylsulfanylpyridin-3-yl)-2,2-dimethyl-propionamide

Hazard: Irritant

Formula: C₁₄H₂₂N₂OS
MW: 266.41 g/mol
MDL Number: MFCD06659005
CAS Number: 766557-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR