2-Chloro-4-iodonicotinic acid

Supplier: Matrix Scientific

026116-500MG
101821-634EA 235.75 USD
101821-634
2-Chloro-4-iodonicotinic acid
2-Chloro-4-iodonicotinic acid

Hazard: Irritant

Formula: C₆H₃ClINO₂
MW: 283.45 g/mol
MDL Number: MFCD05663520
CAS Number: 544671-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR