3-Trimethylsilanylethynylpyridine-2-carboxylic acid amide

Supplier: Matrix Scientific

026110-500MG
101821-622EA 204.62 USD
101821-622
3-Trimethylsilanylethynylpyridine-2-carboxylic acid amide
3-Trimethylsilanylethynylpyridine-2-carboxylic acid amide

Hazard: Irritant

Formula: C₁₁H₁₄N₂OSi
MDL Number: MFCD06658999
CAS Number: 499193-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR