2-Morpholin-4-ylmethylphenylamine

Supplier: Matrix Scientific

Synonyms: 2-(Morpholin-4-ylmethyl)aniline

020824-500MG
101810-836EA 75.38 USD
101810-836
2-Morpholin-4-ylmethylphenylamine
2-Morpholin-4-ylmethylphenylamine

Hazard: Irritant

Formula: C₁₁H₁₆N₂O
MW: 192.26 g/mol
MDL Number: MFCD04114505
CAS Number: 95539-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR