2-(2-Aminoethyl)adamantan-2-ol

Supplier: Matrix Scientific

020823-500MG
101810-834EA 495.38 USD
101810-834
2-(2-Aminoethyl)adamantan-2-ol
2-(2-Aminoethyl)adamantan-2-ol

Hazard: Irritant

Formula: C₁₂H₂₁NO
MDL Number: MFCD08144108
CAS Number: 756818-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR