2-Methyl-2-(4-methylpiperazin-1-yl)propylamine

Supplier: Matrix Scientific

020821-500MG
101810-830EA 280 USD
101810-830
2-Methyl-2-(4-methylpiperazin-1-yl)propylamine
2-Methyl-2-(4-methylpiperazin-1-yl)propylamine

Hazard: Irritant

Formula: C₉H₂₁N₃
MDL Number: MFCD05201608
CAS Number: 21404-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR