2-(3,5-Dimethylpyrazol-1-yl)propylamine dihydrochloride

Supplier: Matrix Scientific

020816-500MG
101810-824EA 280 USD
101810-824
2-(3,5-Dimethylpyrazol-1-yl)propylamine dihydrochloride
2-(3,5-Dimethylpyrazol-1-yl)propylamine dihydrochloride

Hazard: Irritant

Formula: C₈H₁₇Cl₂N₃
MDL Number: MFCD08687687
CAS Number: 1002111-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR