2,5-Dimethylthiophene-3-carboxylic acid
Supplier: Matrix Scientific
020813-500MG
101810-818EA
238.46
USD
101810-818
2,5-Dimethylthiophene-3-carboxylic acid
2,5-Dimethylthiophene-3-carboxylic acid
Hazard: Irritant
Formula:
C₇H₈O₂S MW: 156.2 g/mol |
MDL Number:
MFCD01860238 CAS Number: 26421-32-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...