2-Isopropyl-1H-benzoimidazole-5-carboxylic acid

Supplier: Matrix Scientific

020805-500MG
101810-804EA 280 USD
101810-804
2-Isopropyl-1H-benzoimidazole-5-carboxylic acid
2-Isopropyl-1H-benzoimidazole-5-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₂
MDL Number: MFCD03042800
CAS Number: 505078-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR