3-(4-Methylpiperazin-1-ylmethyl)phenylamine dihydrochloride

Supplier: Matrix Scientific

020803-500MG
101810-800EA 280 USD
101810-800
3-(4-Methylpiperazin-1-ylmethyl)phenylamine dihydrochloride
3-(4-Methylpiperazin-1-ylmethyl)phenylamine dihydrochloride

Hazard: Irritant

Formula: C₁₂H₂₁Cl₂N₃
MDL Number: MFCD08688352
CAS Number: 1185300-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR