4-[3-(4-Fluorophenyl)[1,2,4]oxadiazol-5-yl]butyric acid

Supplier: Matrix Scientific

020791-500MG
101810-778EA 280 USD
101810-778
4-[3-(4-Fluorophenyl)[1,2,4]oxadiazol-5-yl]butyric acid
4-[3-(4-Fluorophenyl)[1,2,4]oxadiazol-5-yl]butyric acid

Hazard: Irritant

Formula: C₁₂H₁₁FN₂O₃
MDL Number: MFCD08133364
CAS Number: 883541-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR