Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Phenyl-1H-pyrazole-4-carboxylic acid

Supplier: Matrix Scientific


020738-500MG
101810-678EA 35.31 USD InStock
101810-678
1-Phenyl-1H-pyrazole-4-carboxylic acid
1-Phenyl-1H-pyrazole-4-carboxylic acid
CAS: 1134-50-5
MDL Number: MFCD00514518
Molecular Formula: C10H8N2O2
Molecular Weight: 188,19
Hazard: Irritant


Formula: C₁₀H₈N₂O₂
MW: 188,19
MDL Number: MFCD00514518
CAS Number: 1134-50-5
EINECS: 000-000-0
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty