5-Methylimidazo[1,2-a]pyridine-2-carboxylic acid

Supplier: Matrix Scientific

020672-500MG
101810-564EA 280 USD
101810-564
5-Methylimidazo[1,2-a]pyridine-2-carboxylic acid
5-Methylimidazo[1,2-a]pyridine-2-carboxylic acid

Hazard: Irritant

Formula: C₉H₈N₂O₂
MW: 176.1720 g/mol
MDL Number: MFCD03419472
CAS Number: 88751-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR