3-Methylisonicotinic acid

Supplier: Matrix Scientific

Synonyms: 3-Methyl-4-pyridinecarboxylic acid

020640-500MG
101810-504EA 148.35 USD
101810-504
3-Methylisonicotinic acid
3-Methylisonicotinic acid

Hazard: Irritant

Formula: C₇H₇NO₂
MW: 137.1360 g/mol
Melting Pt: 235 °C
MDL Number: MFCD00128870
CAS Number: 4021-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR