4-(Cyclopentyloxy)aniline

Supplier: Matrix Scientific

020473-500MG
101810-084EA 261.54 USD
101810-084
4-(Cyclopentyloxy)aniline
4-(Cyclopentyloxy)aniline

Hazard: Irritant

Formula: C₁₁H₁₅NO
MW: 177.2429 g/mol
MDL Number: MFCD00019279
CAS Number: 26455-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR