N-(2-Methoxyethyl)isopropylamine

Supplier: Matrix Scientific

020388-500MG
101809-936EA 75.38 USD
101809-936
N-(2-Methoxyethyl)isopropylamine
N-(2-Methoxyethyl)isopropylamine

CAS: 104678-18-4
MDL Number: MFCD00144828
Molecular Formula: C6H15NO
Molecular Weight: 117,19
Hazard: Irritant

Formula: C₆H₁₅NO
MDL Number: MFCD00144828
CAS Number: 104678-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR