4-Methoxy-3-(2-propynyloxy)benzenecarbaldehyde

Supplier: Matrix Scientific

020340-500MG
101809-844EA 280 USD
101809-844
4-Methoxy-3-(2-propynyloxy)benzenecarbaldehyde
4-Methoxy-3-(2-propynyloxy)benzenecarbaldehyde

Hazard: Irritant

Formula: C₁₁H₁₀O₃
MW: 190.2 g/mol
MDL Number: MFCD03001224
CAS Number: 145654-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR