1,3-Dichloro-2-(2-chloroethoxy)benzene

Supplier: Matrix Scientific

020337-500MG
101809-840EA 495.38 USD
101809-840
1,3-Dichloro-2-(2-chloroethoxy)benzene
1,3-Dichloro-2-(2-chloroethoxy)benzene

Hazard: Irritant

Formula: C₈H₇Cl₃O
MDL Number: MFCD04638771
CAS Number: 53491-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR