2-Bromo-N-(5-methylisoxazol-3-yl)acetamide

Supplier: Matrix Scientific

020323-500MG
101809-812EA 280 USD
101809-812
2-Bromo-N-(5-methylisoxazol-3-yl)acetamide
2-Bromo-N-(5-methylisoxazol-3-yl)acetamide

Hazard: Irritant

Formula: C₆H₇BrN₂O₂
MDL Number: MFCD06194841
CAS Number: 92947-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR