2-Isobutylsulfanylpyrimidine-5-carbaldehyde

Supplier: Matrix Scientific

020318-500MG
101809-802EA 280 USD
101809-802
2-Isobutylsulfanylpyrimidine-5-carbaldehyde
2-Isobutylsulfanylpyrimidine-5-carbaldehyde

Hazard: Irritant

Formula: C₉H₁₂N₂OS
MDL Number: MFCD07801184
CAS Number: 878433-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR