1-Oxo-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

Supplier: Matrix Scientific

020317-500MG
101809-800EA 306.15 USD
101809-800
1-Oxo-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
1-Oxo-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₉NO₃
MDL Number: MFCD00483631
CAS Number: 63586-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR