2-(2-Bromoethoxy)-1,4-dimethylbenzene

Supplier: Matrix Scientific

020311-500MG
101809-788EA 280 USD
101809-788
2-(2-Bromoethoxy)-1,4-dimethylbenzene
2-(2-Bromoethoxy)-1,4-dimethylbenzene

Hazard: Irritant

Formula: C₁₀H₁₃BrO
MDL Number: MFCD02030776
CAS Number: 37136-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR