(2-Pyridin-4-ylthiazol-4-yl)acetic acid

Supplier: Matrix Scientific

020298-500MG
101809-766EA 280 USD
101809-766
(2-Pyridin-4-ylthiazol-4-yl)acetic acid
(2-Pyridin-4-ylthiazol-4-yl)acetic acid

Hazard: Irritant

Formula: C₁₀H₈N₂O₂S
MDL Number: MFCD06739822
CAS Number: 31112-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR