2-(4-Chloro-3,5-dimethylpyrazol-1-yl)ethylamine

Supplier: Matrix Scientific

020294-500MG
101809-758EA 280 USD
101809-758
2-(4-Chloro-3,5-dimethylpyrazol-1-yl)ethylamine
2-(4-Chloro-3,5-dimethylpyrazol-1-yl)ethylamine

Hazard: Irritant

Formula: C₇H₁₂ClN₃
MDL Number: MFCD04967723
CAS Number: 925634-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR