5-Aminomethyl-1,3-dihydrobenzoimidazol-2-one hydrochloride

Supplier: Matrix Scientific

020284-500MG
101809-742EA 607.69 USD
101809-742
5-Aminomethyl-1,3-dihydrobenzoimidazol-2-one hydrochloride
5-Aminomethyl-1,3-dihydrobenzoimidazol-2-one hydrochloride

Hazard: Irritant

Formula: C₈H₁₀ClN₃O
MW: 199.64 g/mol
MDL Number: MFCD13186035
CAS Number: 797809-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR