(1H-Pyrazol-1-yl)acetic acid

Supplier: Matrix Scientific

Synonyms: 1-Pyrazoleacetic Acid

020282-500MG
101809-740EA 395.6 USD
101809-740
(1H-Pyrazol-1-yl)acetic acid
(1H-Pyrazol-1-yl)acetic acid

Hazard: Irritant

Formula: C₅H₆N₂O₂
MW: 126.1133 g/mol
MDL Number: MFCD00228774
CAS Number: 16034-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR