C-(2-Methoxynaphthalen-1-yl)methylamine

Supplier: Matrix Scientific

020276-500MG
101809-730EA 200 USD
101809-730
C-(2-Methoxynaphthalen-1-yl)methylamine
C-(2-Methoxynaphthalen-1-yl)methylamine

Hazard: Irritant

Formula: C₁₂H₁₃NO
MDL Number: MFCD02327725
CAS Number: 136402-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR