3-(4-Fluorophenyl)-1-methyl-1H-pyrazol-5-amine

Supplier: Matrix Scientific

020274-500MG
101809-726EA 209.3 USD
101809-726
3-(4-Fluorophenyl)-1-methyl-1H-pyrazol-5-amine
3-(4-Fluorophenyl)-1-methyl-1H-pyrazol-5-amine

Hazard: Irritant

Formula: C₁₀H₁₀FN₃
MDL Number: MFCD03407433
CAS Number: 126417-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR