Imidazo[1,2-a]pyridine-2-carboxylic acid

Supplier: Matrix Scientific

020265-500MG
101809-712EA 41.54 USD
101809-712
Imidazo[1,2-a]pyridine-2-carboxylic acid
Imidazo[1,2-a]pyridine-2-carboxylic acid

Hazard: Irritant

Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
MDL Number: MFCD03419462
CAS Number: 64951-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR