2-Oxo-6-phenyl-1,2-dihydropyridine-3-carboxylic acid

Supplier: Matrix Scientific

020264-500MG
101809-710EA 484.62 USD
101809-710
2-Oxo-6-phenyl-1,2-dihydropyridine-3-carboxylic acid
2-Oxo-6-phenyl-1,2-dihydropyridine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₉NO₃
MDL Number: MFCD01105052
CAS Number: 56162-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR