Phenoxyacetaldehyde diethyl acetal

Supplier: Matrix Scientific

020252-500MG
101809-694EA 306.15 USD
101809-694
Phenoxyacetaldehyde diethyl acetal
Phenoxyacetaldehyde diethyl acetal

Hazard: Irritant

Formula: C₁₂H₁₈O₃
MW: 210.28 g/mol
MDL Number: MFCD00272279
CAS Number: 32438-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR