4-Methoxy-3-piperidin-1-ylmethylbenzaldehyde

Supplier: Matrix Scientific

020251-500MG
101809-692EA 280 USD
101809-692
4-Methoxy-3-piperidin-1-ylmethylbenzaldehyde
4-Methoxy-3-piperidin-1-ylmethylbenzaldehyde

Hazard: Irritant

Formula: C₁₄H₁₉NO₂
MW: 233.31 g/mol
MDL Number: MFCD01876391
CAS Number: 128501-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR