1-(4-Methylfurazan-3-ylmethyl)piperazine

Supplier: Matrix Scientific

020241-500MG
101809-678EA 280 USD
101809-678
1-(4-Methylfurazan-3-ylmethyl)piperazine
1-(4-Methylfurazan-3-ylmethyl)piperazine

Hazard: Irritant

Formula: C₈H₁₄N₄O
MDL Number: MFCD01346601
CAS Number: 878617-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR