1-Ethyl-2-(3-ethyl-2-methylsulfanyl-3H-imidazol-4-ylmethyl)-1,2,3,4-tetrahydropyrrolo[1,2-a]pyrazin

Supplier: Matrix Scientific

020237-500MG
101809-672EA 280 USD
101809-672
1-Ethyl-2-(3-ethyl-2-methylsulfanyl-3H-imidazol-4-ylmethyl)-1,2,3,4-tetrahydropyrrolo[1,2-a]pyrazin
1-Ethyl-2-(3-ethyl-2-methylsulfanyl-3H-imidazol-4-ylmethyl)-1,2,3,4-tetrahydropyrrolo[1,2-a]pyrazin

Hazard: Irritant

Formula: C₁₆H₂₄N₄S
MDL Number: MFCD08056130
CAS Number: 883539-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR