3-[(Benzo[1,3]dioxol-5-ylmethyl)amino]propan-1-ol

020236-500MG
101809-670EA 197.06 USD
101809-670
3-[(Benzo[1,3]dioxol-5-ylmethyl)amino]propan-1-ol
3-[(Benzo[1,3]dioxol-5-ylmethyl)amino]propan-1-ol

Hazard: Irritant

Formula: C₁₁H₁₅NO₃
MW: 209.25 g/mol
Boiling Pt: 147…151 °C (0.7 Torr)
MDL Number: MFCD04512509
CAS Number: 869943-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR