1-Phenyl-2-[(thiophen-2-ylmethyl)amino]ethanol

Supplier: Matrix Scientific

020235-500MG
101809-668EA 280 USD
101809-668
1-Phenyl-2-[(thiophen-2-ylmethyl)amino]ethanol
1-Phenyl-2-[(thiophen-2-ylmethyl)amino]ethanol

Hazard: Irritant

Formula: C₁₃H₁₅NOS
Melting Pt: 74 °C
MDL Number: MFCD02678520
CAS Number: 66200-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR