Benzo[1,3]dioxol-5-ylmethyl(2-methoxyethyl)amine

Supplier: Matrix Scientific

020234-500MG
101809-666EA 280 USD
101809-666
Benzo[1,3]dioxol-5-ylmethyl(2-methoxyethyl)amine
Benzo[1,3]dioxol-5-ylmethyl(2-methoxyethyl)amine

Hazard: Irritant

Formula: C₁₁H₁₅NO₃
MW: 209.25 g/mol
MDL Number: MFCD07364933
CAS Number: 117986-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR