Methyl[2-(tetrahydropyran-2-ylmethoxy)ethyl]amine

Supplier: Matrix Scientific

020224-500MG
101809-644EA 280 USD
101809-644
Methyl[2-(tetrahydropyran-2-ylmethoxy)ethyl]amine
Methyl[2-(tetrahydropyran-2-ylmethoxy)ethyl]amine

Hazard: Irritant

Formula: C₉H₁₉NO₂
MDL Number: MFCD08146659
CAS Number: 883544-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR