3-Chloro-2-propoxyphenylamine hydrochloride

Supplier: Matrix Scientific

020217-500MG
101809-630EA 280 USD
101809-630
3-Chloro-2-propoxyphenylamine hydrochloride
3-Chloro-2-propoxyphenylamine hydrochloride

Hazard: Irritant

Formula: C₉H₁₃Cl₂NO
MDL Number: MFCD09997047
CAS Number: 1185057-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR