2-(2-Methoxy-1-methylethoxy)phenylamine

Supplier: Matrix Scientific

020214-500MG
101809-624EA 280 USD
101809-624
2-(2-Methoxy-1-methylethoxy)phenylamine
2-(2-Methoxy-1-methylethoxy)phenylamine

Hazard: Irritant

Formula: C₁₀H₁₅NO₂
MDL Number: MFCD08144819
CAS Number: 883545-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR