2-(2-Methoxyethoxy)aniline

Supplier: Matrix Scientific

020212-500MG
101809-620EA 247.69 USD
101809-620
2-(2-Methoxyethoxy)aniline
2-(2-Methoxyethoxy)aniline

Hazard: Irritant

Formula: C₉H₁₃NO₂
MW: 167.2050 g/mol
MDL Number: MFCD08144123
CAS Number: 72806-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR