4-(2-Methoxyethoxy)benzaldehyde

Supplier: Matrix Scientific

020211-500MG
101809-618EA 280 USD
101809-618
4-(2-Methoxyethoxy)benzaldehyde
4-(2-Methoxyethoxy)benzaldehyde

Hazard: Irritant

Formula: C₁₀H₁₂O₃
MDL Number: MFCD08144121
CAS Number: 36824-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR