5-(4-Methoxyphenyl)-2-methyl-2H-pyrazole-3-carboxylic acid

Supplier: Matrix Scientific

020208-500MG
101809-612EA 246.15 USD
101809-612
5-(4-Methoxyphenyl)-2-methyl-2H-pyrazole-3-carboxylic acid
5-(4-Methoxyphenyl)-2-methyl-2H-pyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₃
MW: 232.24 g/mol
MDL Number: MFCD08144799
CAS Number: 93618-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR